5-Bromo-3-chloro-2-fluoro-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

72865-1G 72865-500MG 72865-5G 72865-25G
77472-972EA 415.09 USD
77472-972 77485-590 77496-178 77505-312
5-Bromo-3-chloro-2-fluoro-4-methoxybenzaldehyde ≥97%
5-Bromo-3-chloro-2-fluoro-4-methoxybenzaldehyde

Precaución: For research use only

Formula: C₈H₅BrClFO₂
MW: 267.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764608
CAS Number: 2807469-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR