3-Chloro-5-ethoxy-4-fluorobenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
72816-1G
72816-500MG
72816-5G
77472-968EA
454.23
USD
77472-968
77485-586
77496-174
3-Chloro-5-ethoxy-4-fluorobenzaldehyde ≥97%
3-Chloro-5-ethoxy-4-fluorobenzaldehyde
Precaución: For research use only
Formula:
C₉H₈ClFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD34764579 CAS Number: 2385750-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...