3-(Benzyloxy)-5-chloro-4-fluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

72808-1G 72808-500MG 72808-5G 72808-25G
77472-964EA 415.09 USD
77472-964 77485-582 77496-170 77505-306
3-(Benzyloxy)-5-chloro-4-fluorobenzaldehyde ≥95%
3-(Benzyloxy)-5-chloro-4-fluorobenzaldehyde

Precaución: For research use only

Formula: C₁₄H₁₀ClFO₂
MW: 264.679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764573
CAS Number: 2807471-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR