1-(4-Chloro-3-fluoro-2-methylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

72733-1G 72733-500MG 72733-5G 72733-25G
77472-956EA 415.09 USD
77472-956 77485-574 77496-162 77505-298
1-(4-Chloro-3-fluoro-2-methylphenyl)ethanol ≥95%
1-(4-Chloro-3-fluoro-2-methylphenyl)ethanol

Precaución: For research use only

Formula: C₉H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD34764511
CAS Number: 2901065-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR