1-Bromo-4-chloro-3-(methoxymethoxy)-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

72691-1G 72691-500MG 72691-5G 72691-25G
77472-952EA 415.09 USD
77472-952 77485-570 77496-158 77505-294
1-Bromo-4-chloro-3-(methoxymethoxy)-2-methylbenzene ≥95%
1-Bromo-4-chloro-3-(methoxymethoxy)-2-methylbenzene

Precaución: For research use only

Formula: C₉H₁₀BrClO₂
MW: 265.531 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764492
CAS Number: 2901098-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR