2,2,2-Trifluoro-1-(2-methoxy-4,5-dimethylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

72638-1G 72638-500MG 72638-5G
77472-948EA 683.92 USD
77472-948 77485-566 77496-154
2,2,2-Trifluoro-1-(2-methoxy-4,5-dimethylphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-methoxy-4,5-dimethylphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₁F₃O₂
MW: 232.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07380766
CAS Number: 1369012-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR