3'-(Dimethylamino)-2,2,2-trifluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(3-(Dimethylamino)phenyl)-2,2,2-trifluoroethanone

72627-1G 72627-500MG 72627-5G
77472-944EA 442.9 USD
77472-944 77485-562 77496-150
3'-(Dimethylamino)-2,2,2-trifluoroacetophenone ≥95%
3'-(Dimethylamino)-2,2,2-trifluoroacetophenone

Precaución: For research use only

Formula: C₁₀H₁₀F₃NO
MW: 217.188 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11556187
CAS Number: 33284-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR