2-(Methoxymethyl)-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

72578-1G 72578-500MG 72578-5G 72578-25G
77472-940EA 143.17 USD
77472-940 77485-558 77496-146 77505-286
2-(Methoxymethyl)-5-(trifluoromethyl)benzoic acid ≥97%
2-(Methoxymethyl)-5-(trifluoromethyl)benzoic acid

Precaución: For research use only

Formula: C₁₀H₉F₃O₃
MW: 234.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22492027
CAS Number: 1246522-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR