(4-(4,4-Dimethylcyclohexyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

72563-1G 72563-500MG 72563-5G
77472-936EA 537.66 USD
77472-936 77485-554 77496-142
(4-(4,4-Dimethylcyclohexyl)phenyl)(methyl)sulfane ≥97%
(4-(4,4-Dimethylcyclohexyl)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₅H₂₂S
MW: 234.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764420
CAS Number: 2807440-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR