4-Cyclopentyl-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

72446-1G 72446-500MG 72446-5G
77472-932EA 832.24 USD
77472-932 77485-550 77496-138
4-Cyclopentyl-3-fluorobenzaldehyde ≥97%
4-Cyclopentyl-3-fluorobenzaldehyde

Precaución: For research use only

Formula: C₁₂H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD34764362
CAS Number: 2807445-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR