3,4-Dichloro-2-(methoxymethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

72425-1G 72425-500MG 72425-5G 72425-25G
77472-928EA 415.09 USD
77472-928 77485-546 77496-134 77505-280
3,4-Dichloro-2-(methoxymethoxy)benzaldehyde ≥95%
3,4-Dichloro-2-(methoxymethoxy)benzaldehyde

Precaución: For research use only

Formula: C₉H₈Cl₂O₃
MW: 235.064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764354
CAS Number: 2901096-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR