2-(2-Bromo-4-fluoro-5-methoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

72271-1G 72271-500MG 72271-5G 72271-25G
77472-924EA 415.09 USD
77472-924 77485-542 77496-128 77505-274
2-(2-Bromo-4-fluoro-5-methoxyphenyl)-1,3-dioxolane ≥97%
2-(2-Bromo-4-fluoro-5-methoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₃
MW: 277.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764256
CAS Number: 2901105-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR