1-(3-(1,3-Dioxolan-2-yl)-2-fluoro-5-methoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

72250-1G 72250-500MG 72250-5G 72250-25G
77472-920EA 140.08 USD
77472-920 77485-538 77496-124 77505-272
1-(3-(1,3-Dioxolan-2-yl)-2-fluoro-5-methoxyphenyl)ethanone ≥95%
1-(3-(1,3-Dioxolan-2-yl)-2-fluoro-5-methoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₃FO₄
MW: 240.2276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764243
CAS Number: 2807464-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR