2,2-Difluoro-1-(4-morpholinophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

72222-1G 72222-500MG 72222-5G
77472-916EA 594.31 USD
77472-916 77485-534 77496-120
2,2-Difluoro-1-(4-morpholinophenyl)ethanone ≥95%
2,2-Difluoro-1-(4-morpholinophenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₃F₂NO₂
MW: 241.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28104037
CAS Number: 2173113-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR