1-(4-Bromo-2,5-dimethoxyphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

72108-1G 72108-500MG 72108-5G 72108-25G
77472-912EA 415.09 USD
77472-912 77485-530 77496-116 77505-268
1-(4-Bromo-2,5-dimethoxyphenyl)-2,2-difluoroethanone ≥97%
1-(4-Bromo-2,5-dimethoxyphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₉BrF₂O₃
MW: 295.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764234
CAS Number: 1694272-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR