1-(2,5-Dichloro-3-fluorophenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71974-1G 71974-500MG 71974-5G
77472-908EA 576.8 USD
77472-908 77485-526 77496-112
1-(2,5-Dichloro-3-fluorophenyl)-2,2-difluoroethanone ≥95%
1-(2,5-Dichloro-3-fluorophenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₈H₃Cl₂F₃O
MW: 243.01 g/mol
Storage Temperature: Ambient
CAS Number: 1824062-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR