1-(3-Ethoxy-4-methoxyphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71902-1G 71902-500MG 71902-5G
77472-904EA 474.83 USD
77472-904 77485-522 77496-108
1-(3-Ethoxy-4-methoxyphenyl)-2,2-difluoroethanone ≥95%
1-(3-Ethoxy-4-methoxyphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₁H₁₂F₂O₃
MW: 230.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764214
CAS Number: 2352706-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR