2,2-Difluoro-1-(2-methoxy-4-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71817-1G 71817-500MG 71817-5G
77472-900EA 1008.37 USD
77472-900 77485-518 77496-104
2,2-Difluoro-1-(2-methoxy-4-methylphenyl)ethanone ≥95%
2,2-Difluoro-1-(2-methoxy-4-methylphenyl)ethanone

Precaución: For research use only

Formula: C₁₀H₁₀F₂O₂
MW: 200.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30488929
CAS Number: 2060000-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR