2,2-Difluoro-1-(4-propylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71706-1G 71706-500MG 71706-5G
77472-896EA 594.31 USD
77472-896 77485-514 77496-100
2,2-Difluoro-1-(4-propylphenyl)ethanone ≥95%
2,2-Difluoro-1-(4-propylphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₂F₂O
MW: 198.209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26045124
CAS Number: 1565400-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR