1-(2-Ethoxyphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71704-1G 71704-500MG 71704-5G
77472-892EA 509.85 USD
77472-892 77485-510 77496-096
1-(2-Ethoxyphenyl)-2,2-difluoroethanone ≥95%
1-(2-Ethoxyphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD26045090
CAS Number: 1566808-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR