2,2-Difluoro-1-(2-isopropylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71695-1G 71695-500MG 71695-5G
77472-888EA 576.8 USD
77472-888 77485-506 77496-092
2,2-Difluoro-1-(2-isopropylphenyl)ethanone ≥95%
2,2-Difluoro-1-(2-isopropylphenyl)ethanone

Precaución: For research use only

Formula: C₁₁H₁₂F₂O
MW: 198.209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28103957
CAS Number: 1856798-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR