1-(2,5-Dimethylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71656-1G 71656-500MG 71656-5G
77472-884EA 1037.21 USD
77472-884 77485-502 77496-088
1-(2,5-Dimethylphenyl)-2,2-difluoroethanone ≥95%
1-(2,5-Dimethylphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₁₀F₂O
MW: 184.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20920838
CAS Number: 1335113-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR