1-(2-(Dimethylamino)phenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71642-1G 71642-500MG 71642-5G
77472-880EA 594.31 USD
77472-880 77485-498 77496-084
1-(2-(Dimethylamino)phenyl)-2,2-difluoroethanone ≥95%
1-(2-(Dimethylamino)phenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₀H₁₁F₂NO
Storage Temperature: Ambient
MDL Number: MFCD28103795
CAS Number: 1824052-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR