2,2-Difluoro-1-(4-(methylthio)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

71596-1G 71596-500MG 71596-5G 71596-25G
77472-876EA 381.1 USD
77472-876 77485-494 77496-080 77505-264
2,2-Difluoro-1-(4-(methylthio)phenyl)ethanone ≥95%
2,2-Difluoro-1-(4-(methylthio)phenyl)ethanone

Precaución: For research use only

Formula: C₉H₈F₂OS
MW: 202.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13172969
CAS Number: 81170-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR