5-Bromo-4-chloro-2,3-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

71548-1G 71548-500MG 71548-5G 71548-25G
77472-872EA 415.09 USD
77472-872 77485-490 77496-076 77505-262
5-Bromo-4-chloro-2,3-difluorobenzaldehyde ≥97%
5-Bromo-4-chloro-2,3-difluorobenzaldehyde

Precaución: For research use only

Formula: C₇H₂BrClF₂O
MW: 255.4439 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764163
CAS Number: 2901084-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR