1-(2-(Benzyloxy)-5-methylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

71545-1G 71545-500MG 71545-5G 71545-25G
77472-868EA 415.09 USD
77472-868 77485-486 77496-072 77505-258
1-(2-(Benzyloxy)-5-methylphenyl)-2,2-difluoroethanone ≥95%
1-(2-(Benzyloxy)-5-methylphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₆H₁₄F₂O₂
MW: 276.2779 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34815471
CAS Number: 2921839-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR