(2-(Benzyloxy)-5-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

71540-1G 71540-500MG 71540-5G 71540-25G
77472-864EA 415.09 USD
77472-864 77485-482 77496-068 77505-254
(2-(Benzyloxy)-5-methylphenyl)(methyl)sulfane ≥95%
(2-(Benzyloxy)-5-methylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₅H₁₆OS
MW: 244.3519 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34815469
CAS Number: 2921836-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR