1-(3-Chloro-2-fluoro-5-methoxyphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

71515-1G 71515-500MG 71515-5G 71515-25G
77472-860EA 386.25 USD
77472-860 77485-478 77496-064 77505-250
1-(3-Chloro-2-fluoro-5-methoxyphenyl)ethanol ≥97%
1-(3-Chloro-2-fluoro-5-methoxyphenyl)ethanol

Precaución: For research use only

Formula: C₉H₁₀ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD28527204
CAS Number: 1782494-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR