(5-Bromo-4-fluoro-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

71476-1G 71476-500MG 71476-5G 71476-25G
77472-856EA 220.42 USD
77472-856 77485-474 77496-060 77505-246
(5-Bromo-4-fluoro-2-methylphenyl)(methyl)sulfane ≥97%
(5-Bromo-4-fluoro-2-methylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₈H₈BrFS
Storage Temperature: Ambient
MDL Number: MFCD34764142
CAS Number: 1416465-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR