(5-Iodo-2-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

71427-1G 71427-500MG 71427-5G 71427-25G
77472-852EA 162.74 USD
77472-852 77485-470 77496-056 77505-242
(5-Iodo-2-methoxyphenyl)(methyl)sulfane ≥97%
(5-Iodo-2-methoxyphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₈H₉IOS
Storage Temperature: Ambient
MDL Number: MFCD34764129
CAS Number: 2807441-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR