4-(4-Iodophenyl)tetrahydro-2H-pyran ≥95%

Supplier: AOBChem USA

Warning

71407-1G 71407-500MG 71407-5G 71407-25G
77472-848EA 415.09 USD
77472-848 77485-466 77496-052 77505-238
4-(4-Iodophenyl)tetrahydro-2H-pyran ≥95%
4-(4-Iodophenyl)tetrahydro-2H-pyran

Precaución: For research use only

Formula: C₁₁H₁₃IO
MW: 288.1247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24449763
CAS Number: 1434128-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR