2-Fluoro-6-methoxy-3-(methylthio)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

71366-1G 71366-500MG 71366-5G
77472-844EA 600.49 USD
77472-844 77485-462 77496-048
2-Fluoro-6-methoxy-3-(methylthio)benzonitrile ≥97%
2-Fluoro-6-methoxy-3-(methylthio)benzonitrile

Precaución: For research use only

Formula: C₉H₈FNOS
Storage Temperature: Ambient
MDL Number: MFCD34764104
CAS Number: 1520103-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR