4-(1,3-dioxolan-2-yl)-2,6-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

70828-1G 70828-500MG 70828-5G 70828-25G
77472-840EA 415.09 USD
77472-840 77485-458 77496-044 77505-232
4-(1,3-dioxolan-2-yl)-2,6-difluorobenzaldehyde ≥97%
4-(1,3-dioxolan-2-yl)-2,6-difluorobenzaldehyde

Precaución: For research use only

Formula: C₁₀H₈F₂O₃
MW: 214.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764083
CAS Number: 2445364-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR