3-Chloro-6-ethoxy-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

70561-1G 70561-500MG 70561-5G 70561-25G
77472-836EA 131.84 USD
77472-836 77485-454 77496-038 77505-228
3-Chloro-6-ethoxy-2-fluorobenzaldehyde ≥97%
3-Chloro-6-ethoxy-2-fluorobenzaldehyde

Precaución: For research use only

Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764068
CAS Number: 2385911-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR