6-Fluoro-2-methoxy-3-(methylthio)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

70476-1G 70476-500MG 70476-5G 70476-25G
77472-832EA 379.04 USD
77472-832 77485-450 77496-034 77505-226
6-Fluoro-2-methoxy-3-(methylthio)benzaldehyde ≥95%
6-Fluoro-2-methoxy-3-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₉H₉FO₂S
MW: 200.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764048
CAS Number: 1780785-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR