4-Fluoro-3-formyl-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

70399-1G 70399-500MG 70399-5G
77472-828EA 756.02 USD
77472-828 77485-446 77496-030
4-Fluoro-3-formyl-2-methoxybenzoic acid ≥97%
4-Fluoro-3-formyl-2-methoxybenzoic acid

Precaución: For research use only

Formula: C₉H₇FO₄
MW: 198.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764039
CAS Number: 2166678-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR