2-Fluoro-4-hydroxy-3-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

69673-1G 69673-500MG 69673-5G
77472-824EA 1230.85 USD
77472-824 77485-442 77496-026
2-Fluoro-4-hydroxy-3-methylbenzaldehyde ≥95%
2-Fluoro-4-hydroxy-3-methylbenzaldehyde

Precaución: For research use only

Formula: C₈H₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD22414668
CAS Number: 1824142-12-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR