1-(2-Ethoxy-5-methylphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

69649-1G 69649-500MG 69649-5G
77472-820EA 630.36 USD
77472-820 77485-438 77496-022
1-(2-Ethoxy-5-methylphenyl)ethanol ≥97%
1-(2-Ethoxy-5-methylphenyl)ethanol

Precaución: For research use only

Formula: C₁₁H₁₆O₂
MW: 180.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11053271
CAS Number: 1048917-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR