1-(4-(Benzyloxy)-2,6-dimethylphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

69629-1G 69629-500MG 69629-5G 69629-25G
77472-812EA 415.09 USD
77472-812 77485-432 77496-014 77505-216
1-(4-(Benzyloxy)-2,6-dimethylphenyl)-2,2,2-trifluoroethanone ≥97%
1-(4-(Benzyloxy)-2,6-dimethylphenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₇H₁₅F₃O₂
MW: 308.295 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34815008
CAS Number: 2969365-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR