1-(2,4-Difluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

69492-1G 69492-500MG 69492-5G 69492-25G
77472-808EA 415.09 USD
77472-808 77485-428 77496-008 77505-210
1-(2,4-Difluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥95%
1-(2,4-Difluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₉H₅F₅OS
MW: 256.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34814956
CAS Number: 2969210-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR