(3-Bromo-2,6-difluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

69483-1G 69483-500MG 69483-5G 69483-25G
77472-804EA 82.4 USD
77472-804 77485-424 77496-004 77505-206
(3-Bromo-2,6-difluorophenyl)(methyl)sulfane ≥97%
(3-Bromo-2,6-difluorophenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₇H₅BrF₂S
Storage Temperature: Ambient
MDL Number: MFCD31543063
CAS Number: 2091143-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR