(6-Bromo-2-fluoro-3-propoxyphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

69361-1G 69361-500MG 69361-5G 69361-25G
77472-800EA 415.09 USD
77472-800 77485-420 77496-000 77505-202
(6-Bromo-2-fluoro-3-propoxyphenyl)trimethylsilane ≥97%
(6-Bromo-2-fluoro-3-propoxyphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₂H₁₈BrFOSi
MW: 305.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763912
CAS Number: 2807445-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR