5-Fluoro-2,2'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

69343-1G 69343-500MG 69343-5G 69343-25G
77472-796EA 415.09 USD
77472-796 77485-416 77495-996 77505-198
5-Fluoro-2,2'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
5-Fluoro-2,2'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: For research use only

Formula: C₁₅H₁₃FO
MW: 228.262 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763904
CAS Number: 2901085-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR