(4-Ethoxy-2,6-difluorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

69333-1G 69333-500MG 69333-5G 69333-25G
77472-792EA 229.69 USD
77472-792 77485-412 77495-992 77505-194
(4-Ethoxy-2,6-difluorophenyl)trimethylsilane ≥97%
(4-Ethoxy-2,6-difluorophenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₁H₁₆F₂OSi
Storage Temperature: Ambient
MDL Number: MFCD34763896
CAS Number: 138871-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR