(5-Chloro-2-fluoro-3-(methylthio)phenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

69322-1G 69322-500MG 69322-5G
77472-788EA 601.52 USD
77472-788 77485-408 77495-988
(5-Chloro-2-fluoro-3-(methylthio)phenyl)trimethylsilane ≥97%
(5-Chloro-2-fluoro-3-(methylthio)phenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₀H₁₄ClFSSi
Storage Temperature: Ambient
MDL Number: MFCD34763885
CAS Number: 2901082-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR