(6-Bromo-2-fluoro-3-methoxyphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

69317-1G 69317-500MG 69317-5G 69317-25G
77472-784EA 415.09 USD
77472-784 77485-404 77495-984 77505-188
(6-Bromo-2-fluoro-3-methoxyphenyl)trimethylsilane ≥97%
(6-Bromo-2-fluoro-3-methoxyphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₀H₁₄BrFOSi
MW: 277.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763884
CAS Number: 2918884-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR