1-(4-Ethoxy-2,6-dimethylphenyl)-2,2,2-trifluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
69277-1G
69277-500MG
69277-5G
69277-25G
77472-780EA
339.9
USD
77472-780
77485-400
77495-980
77505-184
1-(4-Ethoxy-2,6-dimethylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(4-Ethoxy-2,6-dimethylphenyl)-2,2,2-trifluoroethanone
Precaución: For research use only
Formula:
C₁₂H₁₃F₃O₂ MW: 246.226 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18375519 CAS Number: 1443354-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...