2-Fluoro-5-methyl-3-(methylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

69273-1G 69273-500MG 69273-5G 69273-25G
77472-776EA 197.76 USD
77472-776 77485-396 77495-976 77505-180
2-Fluoro-5-methyl-3-(methylthio)pyridine ≥97%
2-Fluoro-5-methyl-3-(methylthio)pyridine

Precaución: For research use only

Formula: C₇H₈FNS
MW: 157.209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763864
CAS Number: 2807447-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR