1-(6-Bromo-2-fluoropyridin-3-yl)ethanol ≥97%

Supplier: AOBChem USA

Warning

69250-1G 69250-500MG 69250-5G 69250-25G
77472-772EA 197.76 USD
77472-772 77485-392 77495-972 77505-176
1-(6-Bromo-2-fluoropyridin-3-yl)ethanol ≥97%
1-(6-Bromo-2-fluoropyridin-3-yl)ethanol

Precaución: For research use only

Formula: C₇H₇BrFNO
Storage Temperature: Ambient
MDL Number: MFCD34763854
CAS Number: 2807455-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR