(5-Bromo-2-fluoro-3-propoxyphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

69202-1G 69202-500MG 69202-5G 69202-25G
77472-768EA 416.12 USD
77472-768 77485-388 77495-968 77505-174
(5-Bromo-2-fluoro-3-propoxyphenyl)trimethylsilane ≥97%
(5-Bromo-2-fluoro-3-propoxyphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₂H₁₈BrFOSi
MW: 305.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763837
CAS Number: 2901098-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR